missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) iodide, ultra dry, 99.99% (metals basis)

Product Code. 11386878
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11386878

Brand: Thermo Scientific Alfa Aesar 013619.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light and Air Sensitive and Hygroscopic | CAS: 7783-86-0 | FeI2

Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-86-0
Molecular Formula FeI2
MDL Number MFCD00016094
Synonym iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis

Specifications

Color Gray
Physical Form Beads
Quantity 10 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,diethyl ether and ethanol.
Formula Weight 309.66
Odor Odorless
Sensitivity Air and light sensitive; Hygroscopic
Appearance Gray beads
Density 5.32
Chemical Name or Material Iron(II) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.