missing translation for 'onlineSavingsMsg'
Learn More

Iron Solution for ICP, 1g/L, HCl Matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12963404 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12963404 500 mL 500mL
12943404 100 mL 100mL
12953404 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12963404 Supplier Fisher Chemical Supplier No. S 019 500
  • ONLINE EXCLUSIVE PRICE
    139.00 €
    125.66 € / 500mL
    Save 13.34 €
    9% Off
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Fe
Concentration 10±0.002 g/L
Type Mass Spectrometry Standard
Element Fe
Solvent or Matrix Hydrogen Chloride
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.