missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Product Code. 16391226 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
30 mL
125 mL
250 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
250mL
30mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16391226 30 mL Triple-leached LDPE bottle 30mL
10376122 125 mL Triple-leached LDPE bottle 125mL
10172002 250 mL Triple-leached LDPE bottle 250mL
11448709 500 mL Triple-leached LDPE bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16391226 Supplier SPEX Certiprep Supplier No. PLFE22M
  • ONLINE EXCLUSIVE PRICE
    41.54 €
    38.49 € / 30mL
    Save 3.05 €
    7% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Fe1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 10025-77-1
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Cl3FeH12O6
MDL Number MFCD00149712
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Type Certified Reference Material
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) hexahydrate trichloride
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 270.29
Quantity 30 mL
Chemical Name or Material Iron Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000µg/ml iron
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contain: nitric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.