missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Ionic Strength Adjuster, 2.0M, Reagecon™
BIOPHARMA

Product Code. 11503112 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11503112 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11503112 Supplier Reagecon Supplier No. ISAN5
  • ONLINE EXCLUSIVE PRICE
    229.00 €
    104.47 € / 500mL
    Save 124.53 €
    54% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enables measurement of cations and anions with Ion Selective Electrodes (ISEs). Nitrate Ionic Strength Adjuster, 2.0M, Reagecon™ is added to both samples and ISE standards for use in calibration, control, instrument qualification and method validation in direct and indirect ion measurement.

The use of an Ionic Strength Adjuster (ISA) has several benefits when measuring cations, anions and some dissolved gasses with an Ion Selective Electrode (ISE).

  • The ISA keeps the ionic strength of the sample and standard high, constant and similar, which enables activity measurement to be read as concentration
  • The ISA keeps the pH value constant in some instances, which helps suppress or eliminate interfering ions
  • The ISA eliminates any matrix, hysteresis or erroneous liquid junction potentials that might affect the accuracy of the test result
  • An ISA should never react with the sample or standard chemically

Specifications

CAS 7783-20-2
Density 1.13g/cm3 at 20°C
Physical Form Liquid
Molecular Formula H8N2O4S
Concentration 2M
MDL Number MFCD00003391
Color Colorless
pH 5.8 to 6.2
For Use With (Equipment) Ion Selective Electrode
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name diazanium;sulfate
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
Quantity 500 mL
ChEBI CHEBI:62946
Formula Weight 132.14g/mol
Chemical Name or Material Ammonium sulfate
Show More Show Less
compliance-icons
Product Identifier
  • NITRATE ISE IONIC STRENGTH
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ammonium sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.