missing translation for 'onlineSavingsMsg'
Learn More

Isavuconazole, MedChemExpress

Product Code. 16498627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498627

Brand: Medchem Express HY14273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isavuconazole (BAL-4815) is a triazole prodrug with antifungal activity against yeasts, molds, and dimorphic fungi. Isavuconazole inhibits ergosterol biosynthesis and results in the disruption of fungal membrane structure and function. Isavuconazole is a moderate inhibitor of CYP3A4.

Isavuconazole (BAL-4815) is a triazole prodrug with antifungal activity against yeasts, molds, and dimorphic fungi. Isavuconazole inhibits ergosterol biosynthesis and results in the disruption of fungal membrane structure and function. Isavuconazole is a moderate inhibitor of CYP3A4.

Specifications

Chemical Name or Material Isavuconazole
Physical Form Powder
CAS 241479-67-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H17F2N5OS
For Use With (Application) COVID-19-immunoregulation
Synonym BAL-4815 RO-0094815
Solubility Information DMSO : ≥ 50 mg/mL (114.29 mM)
SMILES FC1=CC=C(F)C=C1[C@](CN2C=NC=N2)(O)[C@@H](C)C3=NC(C4=CC=C(C#N)C=C4)=CS3
Molecular Weight (g/mol) 437.47
Formula Weight 437.47
Percent Purity 99.55%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.