missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Iscove's MDM with 2% FBS

Product Code. 17161036
Encompass
Change view
Click to view available options
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17161036 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161036 Supplier STEMCELL Technologies Supplier No. 07700
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium for preparing and washing samples for colony-forming unit (CFU) assays

  • Iscove's Modified Dulbecco's Medium (MDM) with 2% Fetal Bovine Serum (FBS) is recommended for preparing and washing human cord blood, peripheral blood, or bone marrow samples for colony-forming unit (CFU) assays

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.