missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Iscove's Modified Dulbecco Medium with L-Glutamine

Product Code. 31908429 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908429 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908429 Supplier Sartorius Supplier No. 010581A
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IMDM with- L-Glutamine, without Alpha-Thioglycerol, without Beta-Mercaptoethanol, is a Dulbecco's modification and is considered a more complex and enriched media.

  • IMDM is utilized for specialized cell types and serves as the basis for unique serum-free media formulations.
  • The original formulation demonstrated that erythrocytes and macrophage precursor cells, including primary haemopoietic cells, could be cultured in a defined serum-free medium when supplemented with albumin, lecithin, selenium, and transferrin.
  • IMDM is a modification of DMEM containing selenium, additional amino acids, vitamins, and other components.
  • IMDM is a highly enriched synthetic medium well suited for rapidly proliferating and high-density cell cultures in a 5% CO2 atmosphere.
  • IMDM reportedly supports murine B lymphocytes, haemopoietic tissues from bone marrow, lipopolysaccharide-stimulated B-cells, T-lymphocytes, and a wide variety of hybrid cells.
  • When used with serum, IMDM supports a broad spectrum of mammalian cell types, including bone marrow cells, hybridomas, and lymphocytes.
  • Modified as follows:
    • With L-Glutamine
    • With Phenol Red
    • With Sodium Bicarbonate
    • With HEPES
    • Without Beta Mercaptoethanol
    • Without Alpha-Thioglycerol

Specifications

Form Liquid
Product Type IMDM Cell Culture Media
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.