missing translation for 'onlineSavingsMsg'
Learn More

ISE Buffer Solution Lead, Reagecon™

Product Code. 11748475 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11748475 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11748475 Supplier Reagecon Supplier No. ISEPB5
  • ONLINE EXCLUSIVE PRICE
    310.00 €
    134.80 € / 500mL
    Save 175.20 €
    56% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.