missing translation for 'onlineSavingsMsg'
Learn More

ISO 10382:2002 PCB and Organochlorine Pesticide Mixture 369 100 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30316456
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316456 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316456 Supplier Dr. Ehrenstorfer Supplier No. DREA50000369IO
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣PCB No. 101∣PCB No. 118∣PCB No. 138∣PCB No. 153∣PCB No. 180∣PCB No. 28∣PCB No. 52
CAS 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9, 50-29-3
Analyte or Component Names 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣PCB No. 101∣PCB No. 118∣PCB No. 138∣PCB No. 153∣PCB No. 180∣PCB No. 28∣PCB No. 52
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4'-DDD 100 μg/mL∣2,4'-DDE 100 μg/mL∣2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣Aldrin 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-HCH 100 μg/mL∣Dieldrin 100 μg/mL∣Endrin 100 μg/mL∣gamma-HCH 100 μg/mL∣Heptachlor 100 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 100 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 100 μg/mL∣Hexachlorobenzene 100 μg/mL∣PCB No. 101 100 μg/mL∣PCB No. 118 100 μg/mL∣PCB No. 138 100 μg/mL∣PCB No. 153 100 μg/mL∣PCB No. 180 100 μg/mL∣PCB No. 28 100 μg/mL∣PCB No. 52 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.