missing translation for 'onlineSavingsMsg'
Learn More

ISO 9377-2 Quality Control Mineral Oil Mixture 456 10000 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30313475
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313475 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313475 Supplier Dr. Ehrenstorfer Supplier No. DREV50000456HE
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mineral Oil (without additives)∣No. 2 Diesel Oil
CAS 8042-47-5, 68334-30-5
Analyte or Component Names Mineral Oil (without additives)∣No. 2 Diesel Oil
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix n-Hexane
Industry Type Food Testing
Quantity 5 mL
Concentration or Composition (by Analyte or Components) Mineral Oil (without additives) 10000 μg/mL∣No. 2 Diesel Oil 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.