missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Amyl Acetate, 99+%, Certified AR for Analysis, Mix Of 2- & 3-Methylbutyl Acetate, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
Spécification
Spécification
| Melting Point | -78°C |
| Color | Colorless |
| Boiling Point | 142°C |
| Packaging | Amber glass bottle |
| Quantity | 2.5 L |
| Vapor Density | 4.49 |
| Formula Weight | 130.19g/mol |
| Vapor Pressure | 4mmHg at 20°C |
| Physical Form | Liquid |
Sécurité et traitement
Identifiant produit
- Isoamyl acetate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu