missing translation for 'onlineSavingsMsg'
Learn More

Iso-Amyl Acetate, Extra Pure, SLR, mix of 2- & 3-methylbutyl acetate, Fisher Chemical™

Product Code. 10375500 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10375500

Brand: Fisher Chemical A/6880/17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
MDL Number 8946

Specifications

Melting Point -78°C
Color Colorless
Boiling Point 142°C
Packaging Amber glass bottle
Quantity 2.5 L
Vapor Density 4.49
Formula Weight 130.19g/mol
Vapor Pressure 4mmHg at 20°C
Physical Form Liquid
compliance-icons
Product Identifier
  • Isoamyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.