missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl methacrylate, 85-90%, stabilized

Product Code. 10194540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10194540 250 g Glass bottle 250g
10645711 50 g Glass bottle 50g
10239203 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10194540 Supplier Thermo Scientific Acros Supplier No. 292092500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7534-94-3 | C14H22O2 | 222.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7534-94-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00081070
InChI Key IAXXETNIOYFMLW-UHFFFAOYNA-N
Synonym ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate
PubChem CID 71311141
IUPAC Name [(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 245.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Quantity 250 g
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Stabilizer 100 to 200ppm MEHQ
Formula Weight 222.33
Percent Purity 85-90%
Physical Form Liquid
Chemical Name or Material Isobornyl methacrylate, 85-90%
Show More Show Less
compliance-icons
Product Identifier
  • Isobornyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.