missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl alcohol 5000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30311664
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311664 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311664 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011079ME
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutanol
CAS 78-83-1
Analyte or Component Names Isobutanol
Shipping Condition +20°C
Molecular Formula C4 H10 O
Concentration 5000 μg/mL
Certifications/Compliance ISO 17034
Synonym 1-Propanol, 2-methyl-, Isobutyl alcohol (8CI), 2-Methyl-1-propanol, 1-Hydroxy-2-methylpropane, 1-Isobutanol, 1-Methyl-2-propanol, 2-Methylpropyl alcohol, Isobutanol, Isopropyl carbinol, NSC 5708, iso-Butyl alcohol, 2-Methylpropanol
InChI Formula InChI=1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3
SMILES CC(C)CO
IUPAC Name 2-methylpropan-1-ol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 74.12
Quantity 1 mL
Formula Weight 74.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.