missing translation for 'onlineSavingsMsg'
Learn More

Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10493464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10493464 100 mL AcroSeal™ Glass bottle 100mL
10316494 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10493464 Supplier Thermo Scientific Acros Supplier No. 209521000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5674-02-2 | Molecular Formula: C4H9ClMg | Molecular Weight: 116.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5674-02-2 , 60-29-7
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000471
ChEBI CHEBI:75272

Specifications

Density 0.8300g/mL
Flash Point −040°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H9ClMg
Linear Formula (CH3)2CHCH2MgCl
Quantity 100 mL
Specific Gravity 0.83
Synonym splitomicin, 1,2-dihydro-3h-naphtho 2,1-b pyran-3-one, 1,2-dihydro-3h-benzo f chromen-3-one, 1h-benzo f chromen-3 2h-one, chembl86537, 1,2-dihydrobenzo f chromen-3-one, splitomycin, tocris-1542, d09suo, splitomicin hplc , powder
Solubility Information Solubility in water: reacts.
InChI Key ISFPDBUKMJDAJH-UHFFFAOYSA-N
SMILES C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
IUPAC Name 1,2-dihydrobenzo[f]chromen-3-one
PubChem CID 5269
Formula Weight 116.87
Chemical Name or Material Isobutylmagnesium chloride, 2M solution in diethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutylmagnesium chloride, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.