missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550982
This item is not returnable.
View return policy
Description
GNAT1 Polyclonal Antibody detects endogenous levels of GNAT1 protein.
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phosphodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in rods. This gene is also expressed in other cells, and has been implicated in bitter taste transduction in rat taste cells. Mutations in this gene result in autosomal dominant congenital stationary night blindness. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| GNAT1 | |
| Polyclonal | |
| Unconjugated | |
| GNAT1 | |
| CSNB1G; CSNBAD3; G protein subunit alpha transducin 1; GBT1; Gnat1; Gnat-1; GNATR; guanine nucleotide binding protein (G protein), alpha transducing 1; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; guanine nucleotide binding protein, alpha transducing 1; guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; Ird1; Ird2; irdc; irdr; rod-type transducin alpha subunit; Tr alpha; Tralpha; transducin; transducin alpha subunit; transducin alpha-1 chain; transducin alpha-subunit; transducin, rod-specific; transducin-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14685, 2779, 363143 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11488, P20612 | |
| GNAT1 | |
| Synthesized peptide derived from the Internal region of human Gα t1 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction