missing translation for 'onlineSavingsMsg'
Learn More

Isoeugenol, cis + trans, 98%

Product Code. 11440378
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440378 100 g 100g
11430378 25 g 25g
11450378 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440378 Supplier Thermo Scientific Alfa Aesar Supplier No. B24541.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-54-1 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-54-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009285
InChI Key BJIOGJUNALELMI-ONEGZZNKSA-N
Synonym isoeugenol, e-isoeugenol, trans-isoeugenol, 2-methoxy-4-propenylphenol, 4-propenylguaiacol, 4-hydroxy-3-methoxy-1-propenylbenzene, 2-methoxy-4-prop-1-en-1-yl phenol, trans-p-propenylquaiacol, isoeugenol trans-form, 4-hydroxy-3-methoxypropenylbenzene
PubChem CID 853433
ChEBI CHEBI:50545
IUPAC Name 2-methoxy-4-[(E)-prop-1-enyl]phenol
SMILES COC1=CC(\C=C\C)=CC=C1O

Specifications

Melting Point -10°C
Density 1.08
Boiling Point 266°C to 268°C
Flash Point 134°C (273°F)
Refractive Index 1.577
Quantity 100 g
Beilstein 1909602
Merck Index 14,5171
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Isoeugenol, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.