missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl Acetate, Reagent grade, 98%, Honeywell™

Product Code. 15631040 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
100mL
20 L
500mL
This item is not returnable. View return policy

Product Code. 15631040

Brand: Honeywell Research Chemicals 112674100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00008946
InChI Key MLFHJEHSLIIPHL-UHFFFAOYSA-N
Synonym isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
PubChem CID 31276
ChEBI CHEBI:31725
IUPAC Name 3-methylbutyl acetate
SMILES CC(C)CCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.876g/mL at 25°C
Boiling Point 142°C
Explosion Limit 0.075
Flash Point 25°C (77°F)
Ignition Point 680°F
Packaging Glass Bottle
Linear Formula CH3COOCH2CH2CH(CH3)2
Refractive Index n20/D 1.4(lit.)
Quantity 100 mL
UN Number UN1104
Beilstein 1744750
Vapor Density 4.5 (vs air)
Formula Weight 130.18g/mol
Vapor Pressure 5mmHg (25°C)
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Isopentyl Acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.