missing translation for 'onlineSavingsMsg'
Learn More

Isophorondiamine, 99+%, mixture of cis and trans

Product Code. 10584141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10584141 250 mL Glass bottle 250mL
10727782 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10584141 Supplier Thermo Scientific Supplier No. 352942500
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2855-13-2 | C10H22N2 | 170.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2855-13-2
Molecular Formula C10H22N2
Molecular Weight (g/mol) 170.30
MDL Number MFCD00019397
InChI Key RNLHGQLZWXBQNY-UHFFFAOYNA-N
Synonym isophorone diamine, isophoronediamine, 3-aminomethyl-3,5,5-trimethylcyclohexylamine, 3-aminomethyl-3,5,5-trimethylcyclohexanamine, cyclohexanemethanamine, 5-amino-1,3,3-trimethyl, ipda, ccris 6680, 5-amino-1,3,3-trimethylcyclohexanemethylamine, cyclohexanemethylamine, 5-amino-1,3,3-trimethyl, 1-amino-3-aminomethyl-3,5,5-trimethylcyclohexane
PubChem CID 17857
IUPAC Name 3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
SMILES CC1(C)CC(N)CC(C)(CN)C1

Specifications

Melting Point 10°C
Density 0.9200g/mL
Boiling Point 247°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4880 to 1.49
Quantity 250 mL
Specific Gravity 0.92
Solubility Information Solubility in water: miscible with water
Viscosity 18 mPa.s (20°C)
Formula Weight 170.3
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Isophorondiamine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isophorondiamine, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.