missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isoprenaline hemisulfate

Product Code. p-7112903 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15773457

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299-95-6 | C11H17NO3·1/2H2O4S | 520.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-95-6
Molecular Formula C11H17NO3·1/2H2O4S
Molecular Weight (g/mol) 520.59
InChI Key ZOLBALGTFCCTJF-UHFFFAOYSA-N
Synonym isoprenaline sulphate, isoprenaline sulfate, aleudrin, novodrine, aleudrine sulfate, isoproterenol sulfate, dl-isoprenaline sulfate, dl-isoproterenol sulfate, d,l-isoproterenol sulfate, +--isoprenaline sulfate
PubChem CID 8239
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid
SMILES CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O

Specifications

Melting Point 83.0°C
Quantity 5 g
Formula Weight 520.59
Chemical Name or Material Isoprenaline hemisulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.