missing translation for 'onlineSavingsMsg'
Learn More

Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11925431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925431 100 g 100g
11683009 5 g 5g
11915431 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925431 Supplier Thermo Scientific Acros Supplier No. 437211000
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-30-9
Molecular Formula C11H18ClNO3
Molecular Weight (g/mol) 247.72
MDL Number MFCD00012603,MFCD00064548
InChI Key IROWCYIEJAOFOW-UHFFFAOYNA-N
Synonym isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
PubChem CID 5807
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 165.0°C to 175.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: soluble in cold water.
Formula Weight 247.72
Percent Purity 98%
Chemical Name or Material Isoprenaline hydrochloride
compliance-icons
Product Identifier
  • Isoprenaline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Isoprenaline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.