missing translation for 'onlineSavingsMsg'
Learn More

Isoprenaline hydrochloride, MedChemExpress

Product Code. 16472189
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472189 1 g 1g
16441426 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472189 Supplier Medchem Express Supplier No. HYB0468
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isoprenaline hydrochloride is a non-selective β-adrenergic receptor agonist with potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities.

Isoprenaline hydrochloride is a non-selective β-adrenergic receptor agonist with potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities.

Specifications

Chemical Name or Material Isoprenaline hydrochloride
CAS 51-30-9
Color White
Purity Grade Notes Research
Molecular Formula C11H18ClNO3
Quantity 1 g
Synonym Isoproterenol hydrochloride
Solubility Information DMSO : 80 mg/mL (322.95 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (201.84 mM)
SMILES OC1=CC=C(C(O)CNC(C)C)C=C1O.Cl
Molecular Weight (g/mol) 247.72
Formula Weight 247.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.