missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol-d8, for NMR, 99+ atom % D

Product Code. p-151151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10499310 5 g Glass Bottle 5g
10571431 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10499310 Supplier Thermo Scientific Acros Supplier No. 174850050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 22739-76-0 | C3H8O | 68.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22739-76-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 68.15
MDL Number MFCD00044341
InChI Key KFZMGEQAYNKOFK-PIODKIDGSA-N
Synonym 2-propanol-d8, isopropanol-d8, 2 h? propan-2-2 h ol, octadeuteroisopropanol, 2h8-2-propanol, 2-propan-1,1,1,2,3,3,3-d7-ol-d, 2-propanol-d isotopic 1g, 2-propanol-d8, 99.5 atom % d
PubChem CID 2723972
IUPAC Name 1,1,1,2,3,3,3-heptadeuterio-2-deuteriooxypropane
SMILES [2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 0.8900g/mL
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Quantity 5 g
Linear Formula (CD3)2CDOD
Formula Weight 68.15
Grade NMR
Chemical Name or Material Isopropanol-d8, 99+ atom % D
compliance-icons compliance-icons
Product Identifier
  • Isopropanol-d8
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.