missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-hydroxybenzoate, 98%

Product Code. 11498860
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498860 5 g 5g
11431658 10 g 10g
11441658 50 g 50g
11451658 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498860 Supplier Thermo Scientific Alfa Aesar Supplier No. A13930.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4191-73-5 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as a preservative in cosmetics.

Solubility
Soluble in Chloroform, Ethyl Acetate.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016468
InChI Key CMHMMKSPYOOVGI-UHFFFAOYSA-N
Synonym isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
PubChem CID 20161
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 84°C to 86°C
Quantity 5 g
Beilstein 2641564
Solubility Information Soluble in Chloroform,Ethyl Acetate.
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Isopropyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.