missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Paraben, TRC

Product Code. 17924275
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924275 5 g 5g
17901675 10 g 10g
17953765 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924275 Supplier Toronto Research Chemicals Supplier No. TRCI8728605G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym Benzoic acid, 4-hydroxy-, 1-methylethyl ester, Benzoic acid, p-hydroxy-, isopropyl ester (6CI,7CI,8CI), 4-Hydroxybenzoic acid isopropyl ester, Isopropyl 4-hydroxybenzoate, Isopropyl p-hydroxybenzoate, Isopropylparaben, p-Hydroxybenzoic acid isopropyl ester
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)c1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12O3/c1-7(2)13-10(12)8-3-5-9(11)6-4-8/h3-7,11H,1-2H3
Formula Weight 180.0786
Percent Purity >95%
Chemical Name or Material Isopropyl Paraben
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.