missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl salicylate, 99%

Product Code. p-152080 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 10001811

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isopropyl salicylate, 99%, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 122.0 deg.C (18.0 mmHg), Colorless to Yellow, Molecular Weight: 180.203, Percent Purity: 99%, Assay Percent Range: 99%, CAS: 607-85-2, ChEBI: CHEBI:38703, Density: 1.0600g/mL, EINECS Number: 210-143-2 | CAS: 607-85-2 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-85-2
MDL Number MFCD00035703
InChI Key YEULQIJMIOWCHB-UHFFFAOYSA-N
Synonym isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
PubChem CID 11838
ChEBI CHEBI:38703
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1O

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 122.0°C (18.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H12O3
Refractive Index 1.5090 to 1.5110
Quantity 100 g
Specific Gravity 1.06
Molecular Weight (g/mol) 180.2
Formula Weight 180.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl salicylate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Isopropyl salicylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.