missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Salicylate, TRC

Product Code. 30133601
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133601 25 g 25g
30132481 100 g 100g
17940845 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133601 Supplier Toronto Research Chemicals Supplier No. TRCI87297525G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-85-2
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym 2-Hydroxybenzoic acid 1-methylethyl ester, Salicylic acid, isopropyl ester (6CI,7CI,8CI), 2-Hydroxybenzoic acid isopropyl ester, Isopropyl o-hydroxybenzoate, Isopropyl salicylate
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)c1ccccc1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H12O3/c1-7(2)13-10(12)8-5-3-4-6-9(8)11/h3-7,11H,1-2H3
Formula Weight 180.0786
Chemical Name or Material Isopropyl Salicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.