missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-1-carbonitrile, 99%

Product Code. 11348726
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11348726 1 g 1g
11358726 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11348726

Brand: Thermo Scientific Alfa Aesar L19425.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1198-30-7 | C10H6N2 | 154.172 g/mol

Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198-30-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00134166
InChI Key HJHXYSBRTVFEDD-UHFFFAOYSA-N
Synonym 1-cyanoisoquinoline, 1-isoquinolinecarbonitrile, isoquinaldonitrile, isoquinolinecarbonitrile, 1-cyano-isoquinoline, isoquinaldinic nitrile, pubchem6269, maybridge1_002114, timtec-bb sbb002487
PubChem CID 306057
IUPAC Name isoquinoline-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CN=C2C#N

Specifications

Melting Point 87°C to 91°C
Odor Odorless
Quantity 1 g
Beilstein 119104
Solubility Information Insoluble in water.
Formula Weight 154.17
Percent Purity 99%
Chemical Name or Material Isoquinoline-1-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.