missing translation for 'onlineSavingsMsg'
Learn More

Isovaleramide, 98%

Product Code. 11312866
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312866 5 g 5g
11322866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312866 Supplier Thermo Scientific Alfa Aesar Supplier No. L09072.06
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-46-8 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-46-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014807
InChI Key SANOUVWGPVYVAV-UHFFFAOYSA-N
Synonym isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
PubChem CID 10930
IUPAC Name 3-methylbutanamide
SMILES CC(C)CC(=O)N

Specifications

Melting Point 134°C to 138°C
Boiling Point 232°C
Quantity 5 g
Beilstein 1740789
Merck Index 14,5230
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material Isovaleramide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.