missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric anhydride, 95%

Product Code. 11326926
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326926 5 g 5g
11336926 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326926 Supplier Thermo Scientific Alfa Aesar Supplier No. L04385.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1468-39-9 | C10H18O3 | 186.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-39-9
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00015051
InChI Key FREZLSIGWNCSOQ-UHFFFAOYSA-N
Synonym isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
PubChem CID 73847
IUPAC Name 3-methylbutanoyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC(=O)CC(C)C

Specifications

Melting Point -65°C
Density 0.927
Boiling Point 103°C to 104°C (24 mmHg)
Flash Point 91°C (195°F)
Linear Formula [(CH3)2CHCH2CO]2O
Refractive Index 1.4165
Quantity 5 g
UN Number UN3265
Beilstein 1635850
Sensitivity Moisture sensitive
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Isovaleric anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.