missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITFG1 Monoclonal Antibody (936213)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561979
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Modulator of T-cell function. Has a protective effect in graft versus host disease model.
Specifications
| ITFG1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8TB96 | |
| Itfg1 | |
| Human embryonic kidney cell line HEK293-derived human ITFG1, Met1-Ile566. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 936213 | |
| Unconjugated | |
| Itfg1 | |
| 2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 81533 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction