missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITGA5 Monoclonal Antibody (5H10.27), FITC
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ A15400

Product Code. 15265714

  • 462.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ITGA5 Monoclonal Antibody for Flow

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITGA5
Monoclonal
0.5 mg/mL
PBS with <0.1% sodium azide; pH 7.4
P11688
Itga5
C57BL/6 x A/J F1 mouse mast cell line MC/9 1.
100 μg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry
5H10.27
FITC
Itga5
2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A
Rat
Purified
RUO
16402
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.