missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
275.00 € - 565.00 €
Specifications
| Antigen | ITIH2 |
|---|---|
| Dilution | Western Blot Reported in scientific literature (PMID: 30623115)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen Reported in scientific literature (PMID: 29855321). |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451952
|
Novus Biologicals
NBP2-31750-25ul |
25 μL |
275.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18113983
|
Novus Biologicals
NBP2-31750 |
0.1 mL |
565.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITIH2 Polyclonal specifically detects ITIH2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| ITIH2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| H2P, IGHEP2, inter-alpha (globulin) inhibitor H2, inter-alpha (globulin) inhibitor, H2 polypeptide, Inter-alpha-inhibitor heavy chain 2, Inter-alpha-trypsin inhibitor complex component II, inter-alpha-trypsin inhibitor heavy chain H2, ITI heavy chain H2, ITI-HC2, Serum-derived hyaluronan-associated protein, SHAP | |
| ITIH2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot Reported in scientific literature (PMID: 30623115)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen Reported in scientific literature (PMID: 29855321). | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P19823 | |
| 3698 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LPGAKVQFELHYQEVKWRKLGSYEHRIYLQPGRLAKHLEVDVWVIEPQGLRFLHVPDTFEGHFDGVPVISKGQQKAHVSFKPTVAQQRICPNCRET | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title