missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITLN1 Monoclonal Antibody (746420)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561904

Product Code. 30535559

  • 469.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITLN1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O88310
ITLN1
E. coli-derived recombinant mouse Intelectin-1/Omentin, Ala20-Ser298.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
746420
Unconjugated
ITLN1
calcium-dependent galactosyl-specific lectin; endothelial lectin HL-1; FLJ20022; Galactofuranose-binding lectin; hIntL; HL1; HL-1; intelectin 1; intelectin 1 (galactofuranose binding); intelectin 1 (galactofuranose binding)-like; intelectin 2; intelectin 5; intelectin a; intelectin-1; Intelectin-1a; intestinal lactoferrin receptor; INTL; Itln; ITLN1; ITLN-1; Itln1a; Itln2; Itln5; Itlna; LFR; omentin; UNQ640/PRO1270
Rat
Protein A/G
RUO
16429
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.