missing translation for 'onlineSavingsMsg'
Learn More

Itopride hydrochloride, MedChemExpress

Product Code. 16462206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462206 500 mg 500mg
16441048 5 g 5g
16438436 100 mg 100mg
16422269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462206 Supplier Medchem Express Supplier No. HYB0732
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Itopride (HSR803) hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. Itopride hydrochloride enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, can be used as a gastrointestinal prokinetic agent. Itopride can be used for researching gastro-esophageal reflux disease (GERD).

Itopride (HSR803) hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. Itopride hydrochloride enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, can be used as a gastrointestinal prokinetic agent. Itopride can be used for researching gastro-esophageal reflux disease (GERD).

Specifications

Chemical Name or Material Itopride hydrochloride
CAS 122892-31-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H27ClN2O4
Quantity 500 mg
Synonym HSR803
Solubility Information DMSO : 100 mg/mL (253.24 mM; Need ultrasonic) ∣H2O : 50 mg/mL (126.62 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=C(OCCN(C)C)C=C1)C2=CC=C(OC)C(OC)=C2.Cl
Molecular Weight (g/mol) 394.89
Formula Weight 394.89
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.