missing translation for 'onlineSavingsMsg'
Learn More

Itopride Hydrochloride, TRC

Product Code. 30010415
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010415 50 mg 50mg
17944085 10 mg 10mg
17922865 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010415 Supplier Toronto Research Chemicals Supplier No. TRCI93150050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 122892-31-3
Molecular Formula C20 H26 N2 O4 . Cl H
Molecular Weight (g/mol) 394.89
Synonym N-[[4-[2-(Dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxybenzamide Hydrochloride (1:1), Benzamide, N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxy-, monohydrochloride (9CI), Ganaton, HSR 803, Itopride hydrochloride
IUPAC Name N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxybenzamide;hydrochloride
SMILES Cl.COc1ccc(cc1OC)C(=O)NCc2ccc(OCCN(C)C)cc2

Specifikationer

Quantity 50 mg
InChI Formula InChI=1S/C20H26N2O4.ClH/c1-22(2)11-12-26-17-8-5-15(6-9-17)14-21-20(23)16-7-10-18(24-3)19(13-16)25-4;/h5-10,13H,11-12,14H2,1-4H3,(H,21,23);1H
Formula Weight 394.1659
Percent Purity >95%
Chemical Name or Material Itopride Hydrochloride
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.