missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPA Recombinant Rabbit Monoclonal Antibody (23GB5975), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | ITPA |
|---|---|
| Clone | 23GB5975 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088227
![]() |
Invitrogen™
MA552962 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089188
|
Invitrogen™
MA552963 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITPA Recombinant Monoclonal antibody specifically detects ITPA in Human, Mouse samples. It is validated for Western BlotSpecifications
| ITPA | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9BY32, Q9D892 | |
| 16434, 3704 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5975 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2010016I08Rik; AU020102; C20orf37; dJ794I6.3; HAMAP-Rule:MF_03148}; HLC14-06-P; inosine triphosphatase; inosine triphosphatase (nucleoside triphosphate pyrophosphatase); inosine triphosphatase {ECO:0000255; inosine triphosphate pyrophosphatase; inosine triphosphate pyrophosphatase {ECO:0000255; inosine triphosphate pyrophosphohydrolase; Itp; ITPA; ITPase; ITPase {ECO:0000255; My049; My049 protein; Non-canonical purine NTP pyrophosphatase; non-canonical purine NTP pyrophosphatase {ECO:0000255; non-standard purine NTP pyrophosphatase; non-standard purine NTP pyrophosphatase {ECO:0000255; NTPase; NTPase {ECO:0000255; nucleoside triphosphate pyrophosphatase; nucleoside-triphosphate diphosphatase; nucleoside-triphosphate diphosphatase {ECO:0000255; Nucleoside-triphosphate pyrophosphatase; nucleoside-triphosphate pyrophosphatase {ECO:0000255; OK/SW-cl.9; Putative oncogene protein hlc14-06-p | |
| A synthesized peptide derived from human ITPA (111-143AA). | |
| Primary | |
| -20°C | |
| ITPA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title