missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38522-100ul
This item is not returnable.
View return policy
Description
ITPK1 Polyclonal antibody specifically detects ITPK1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ITPK1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.7.1.134, EC 2.7.1.159, inositol 13,4-triphosphate 5/6 kinase, inositol 13,4-trisphosphate 5/6 kinase, Inositol 13,4-trisphosphate 5/6-kinase, inositol-tetrakisphosphate 1-kinase, Inositol-triphosphate 5/6-kinase, Ins(13,4)P(3) 5/6-kinase, ITRPK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 185-414 of human ITPK1 (NP_055031.2).,, Sequence:, CVVQNFINHNAVLYKVFVVGESYTVVQRPSLKNFSAGTSDRESIFFNSHNVSKPESSSVLTELDKIEGVFERPSDEVIRELSRALRQALGVSLFGIDIIINNQTGQHAVIDINAFPGYEGVSEFFTDLLNHIATVLQGQSTAMAATGDVALLRHSKLLAEPAGGLVGERTCSASPGCCGSMMGQDAPWKAEADAGGTAKLPHQRLGCNAGVSPSFQQHCVASLATKASSQ | |
| 100 μL | |
| Signal Transduction | |
| 3705 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction