missing translation for 'onlineSavingsMsg'
Learn More

IVD Antibody (04), Novus Biologicals™

Mouse Monoclonal Antibody

220.00 € - 326.00 €

Specifications

Antigen IVD
Clone 04
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.1-0.5 uL/mg of lysate
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18689816
View Documents
Novus Biologicals
NBP3-06536-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18623407
View Documents
Novus Biologicals
NBP3-06536-50ul
50 μg
220.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IVD Monoclonal antibody specifically detects IVD in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications

Specifications

IVD
Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.1-0.5 uL/mg of lysate
Monoclonal
Purified
RUO
Human
ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IVD (Accession#: AAH17202.1; His30-His423).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
04
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Stem Cell Markers
0.2 um filtered solution in PBS
3712
IgG2b
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.