missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin, ≥95%, MP Biomedicals™

Product Code. 11412651 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412651 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412651 Supplier MP Biomedicals Supplier No. 0219600991
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    129.15 € / 1g
    Save 7.85 €
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ivermectin is a broad-spectrum antiparasitic agent. Ivermectin is a macrolide antiobiotic, & semisynthetic derivative of avermectin B1a and B1b

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
MDL Number MFCD00869511
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym 22, 23-Dihydroavermectin B1, Hyvermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Type Ivermectin
Physical Form Crystalline Powder
Color White or White/Yellow
Quantity 1 g
Assay Percent Range ≥95%
Formula Weight 875.1
Percent Purity ≥95%
Grade Not Available
Chemical Name or Material Ivermectin
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ivermectin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ivermectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.