missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin, MedChemExpress

Product Code. 16471766
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471766 500 mg 500mg
16471499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471766 Supplier Medchem Express Supplier No. HY15310
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ivermectin (MK-933) is a broad-spectrum anti-parasite agent.

Ivermectin (MK-933) is a specific inhibitor of Impα/β1-mediated nuclear import and has potent antiviral activity towards both HIV-1 and dengue virus. It is a positive allosteric effector of P2X4 and the α7 neuronal nicotinic acetylcholine receptor (nAChRs). Ivermectin also inhibits bovine herpesvirus1 (BoHV-1) replication and inhibits BoHV-1 DNA polymerase nuclear import. Ivermectin is a candidate therapeutic against SARS-CoV-2/COVID-19.

Specifications

Chemical Name or Material Ivermectin
Physical Form Solid
CAS 70288-86-7
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C48H74O14
For Use With (Application) COVID-19-anti-virus
Synonym MK-933
Solubility Information DMSO : 250 mg/mL (285.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@@H]3C[C@H]4C[C@@]5(O[C@]([H])(C(C)C)[C@@H](C)CC5)O3)[C@@H](C)/C=C/C=C6CO[C@@]7([H])[C@]\6(O)[C@H](C(O4)=O)C=C(C)[C@H]7O)C[C@@H]2OC)O[C@@H](C)[C@@H]1O.CO[C@H]8C[C@H](O[C@H]9[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@@H]%10C[C@H]%11C[C@@]%12(O[C@]([H])([C@@H](C)CC)[C@@H](C)CC%12)O%10)[C@@H](C)/C=C/C=C%13CO[C@@]%14([H])[C@]\%13(O)[C@H](C(O%11)=O)C=C(C)[C@H]%14O)C[C@@H]9OC)O[C@@H](C)[C@@H]8O
Molecular Weight (g/mol) 875.09
Formula Weight 875.09
Percent Purity 96.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.