missing translation for 'onlineSavingsMsg'
Learn More

IWP-2, MedChemExpress

Product Code. 16495908
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495908 10 mg 10mg
16482057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495908 Supplier Medchem Express Supplier No. HY13912
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress IWP-2 is an inhibitor of Wnt processing and secretion with an IC50 of 27 nM. IWP-2 targets the membrane-bound O-acyltransferase porcupine (Porcn) and thus preventing a crucial Wnt ligand palmitoylation. IWP-2 is also an ATP-competitive CK1δ inhibitor with an IC50 of 40 nM for the gatekeeper mutant M82FCK1δ.

IWP-2 is an inhibitor of Wnt processing and secretion with an IC50 of 27 nM. IWP-2 targets the membrane-bound O-acyltransferase porcupine (Porcn) and thus preventing a crucial Wnt ligand palmitoylation. IWP-2 is also an ATP-competitive CK1δ inhibitor with an IC50 of 40 nM for the gatekeeper mutant M82FCK1δ.

Specifications

Chemical Name or Material IWP-2
Physical Form Powder
CAS 686770-61-6
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H18N4O2S3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMF : 12.5 mg/mL (26.79 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 2 mg/mL (4.29 mM; Need ultrasonic)
SMILES O=C(NC1=NC2=CC=C(C)C=C2S1)CSC3=NC(CCS4)=C4C(N3C5=CC=CC=C5)=O
Molecular Weight (g/mol) 466.6
Formula Weight 466.6
Percent Purity 95.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.