missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IZUMO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69692
This item is not returnable.
View return policy
Description
IZUMO1 Polyclonal specifically detects IZUMO1 in Human samples. It is validated for Western Blot.
Specifications
| IZUMO1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8IYV9 | |
| IZUMO1 | |
| Synthetic peptides corresponding to IZUMO1(izumo sperm-egg fusion 1) The peptide sequence was selected from the C terminal of IZUMO1. Peptide sequence SLALITGLTFAIFRRRKVIDFIKSSLFGLGSGAAEQTQVPKEKATDSRQQ. | |
| Affinity purified | |
| RUO | |
| 284359 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction