missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAB1 Antibody (8H8.5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB200-312
This item is not returnable.
View return policy
Description
JAB1 Monoclonal specifically detects JAB1 in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| JAB1 | |
| Monoclonal | |
| 1mg/mL | |
| Western Blot, Immunoprecipitation | |
| COPS5 | |
| The full-length mouse JAB-1 gene expressed in E. coli. | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for JAB-1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 8H8.5 | |
| Unconjugated | |
| COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5 | |
| Mouse | |
| 38 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 10987 | |
| Human, Mouse, Drosophila | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction