missing translation for 'onlineSavingsMsg'
Learn More

JLK 6, TRC

Product Code. 30389172
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389172 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389172 Supplier Toronto Research Chemicals Supplier No. TRCJ21198525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material JLK 6
CAS 62252-26-0
Molecular Formula C10H8ClNO3
Quantity 25 mg
InChI Formula InChI=1S/C10H8ClNO3/c1-14-10-8(11)6-3-2-5(12)4-7(6)9(13)15-10/h2-4H,12H2,1H3
SMILES COC1=C(Cl)c2ccc(N)cc2C(=O)O1
IUPAC Name 7-amino-4-chloro-3-methoxyisochromen-1-one
Molecular Weight (g/mol) 225.63
Formula Weight 225.0193 g/mol
Percent Purity >95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.