missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77074-0.025mg
This item is not returnable.
View return policy
Description
JMJD5 Polyclonal specifically detects JMJD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| JMJD5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 1.14.11.27, FLJ13798, JmjC domain-containing protein 5, jumonji domain containing 5, Jumonji domain-containing protein 5, KDM8, lysine-specific demethylase 8 | |
| Rabbit | |
| 47.3 kDa | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Q8N371 | |
| KDM8 | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human JMJD5. The immunogen is located within amino acids 350 - 400 of JMJD5. Amino Acid Squence: TSQVDVENPDLEKFPKFA | |
| Affinity Purified | |
| RUO | |
| 79831 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction