missing translation for 'onlineSavingsMsg'
Learn More

JNJ 10191584 maleate, Thermo Scientific™

Product Code. 15466899
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466899 50 mg 50mg
15456899 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466899 Supplier Thermo Scientific Alfa Aesar Supplier No. J65433.MF
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A highly selective histamine H4 receptor silent antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869497-75-6
Molecular Formula C17H21ClN4O5
Molecular Weight (g/mol) 396.83
MDL Number MFCD09878266
InChI Key BODOIXYJOORQBQ-UHFFFAOYSA-N
Synonym jnj 10191584 maleate
PubChem CID 77519402
IUPAC Name (6-chloro-1H-benzimidazol-2-yl)-(4-methylpiperazin-1-yl)methanone;formic acid;prop-2-enoic acid
SMILES OC=O.OC(=O)C=C.CN1CCN(CC1)C(=O)C1=NC2=CC=C(Cl)C=C2N1

Specifications

Color Tan to White
Quantity 50 mg
Formula Weight 394.81
Physical Form Powder
Chemical Name or Material JNJ 10191584 maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.