missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JNJ 10191584 maleate
GREENER_CHOICE

Product Code. 18075572
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075572 50 mg 50mg
18044202 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075572 Supplier Tocris Bioscience™ Supplier No. 2441/50
  • ONLINE EXCLUSIVE PRICE
    816.00 €
    771.75 € / 50mg
    Save 44.25 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective H4 receptor antagonist; orally active

Highly selective histamine H4 receptor silent antagonist; binds with high affinity to the human H4 receptor (Ki = 26 nM). > 540-fold selective over the H3 receptor (Ki = 14.1 μM). Inhibits mast cell and eosinophil chemotaxis in vitro with IC50 values of 138 and 530 nM respectively. Orally active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869497-75-6
Molecular Formula C17H21ClN4O5

Specifications

Quantity 50 mg
Formula Weight 394.81
Purity >99%
Molecular Weight (g/mol) 396.83
Chemical Name or Material JNJ 10191584 maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.