missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JNK1 Recombinant Rabbit Monoclonal Antibody (K01_2T92)

Product Code. 30830350
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830350 50 μL
30824752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830350 Supplier Invitrogen™ Supplier No. MA565733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

JNK1 Recombinant Monoclonal Antibody for Western Blot

MAPK8 (JNK1) is activated by the presence of misfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity. It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2T92
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene MAPK8
Gene Accession No. P45983, P49185, Q91Y86
Gene Alias AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; zgc:112379
Gene Symbols MAPK8
Host Species Rabbit
Immunogen Recombinant protein of human JNK1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100771734, 116554, 26419, 5599
Target Species Human, Mouse, Rat, Hamster
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.