Learn More
3M™ Jupiter™ Starter Kits
Product Code. 11741403
Click to view available options
Filter Type:
OV/HE Filter
Unit Size:
Each
Product Code. 11741403
Brand: 3M™ 7000079735
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ready-to-use starter kits feature everything needed for filtration in the laboratory. Complete with electronic audible and visual alarms in the event of low battery and/or low air flow.
  • High comfort decontamination belt
  • Air Flow Indicator and Calibration tube
  • Pair of Organic Vapour and Particulate (A2P) Filters and Prefilters
  • Length-adjusting breathing tube
  • 8-hour rechargeable battery (NiMH) and a battery charger
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 8 hours
Battery Type NiMH
Filter Type OV/HE Filter
Product Type PAPR Assembly, No Headtop
For Use With (Application) Protection against gases, vapors, and particulates, or a combination of contaminants per filter used
Includes Powered turbo unit, decontamination belt, calibration tube and airflow indicator, 2 x A2P filters, pre-filters, breathing tube, rechargeable battery, battery charger
Model 56480
Quantity 1 Ea.